To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Ribosomal Protein L17
- Antigen Symbol:RPL17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:6139
- Antigen Synonyms:PD-1|FLJ18762,60S ribosomal protein L23|FLJ92089|rpL23|MGC117162|ribosomal protein L17|RPL23,60S ribosomal protein L17|gene encoding putative NFkB activating protein
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence RYSLDPENPTKSC corresponding to N-Terminus according to NP_001030178.1.
- Tested Applications:Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102154-678
- Supplier no.:NBP1-00237
Specifications
About this item
The Ribosomal Protein L17 Antibody from Novus Biologicals is a goat polyclonal antibody to Ribosomal Protein L17. This antibody reacts with human. The Ribosomal Protein L17 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Type: Primary
Antigen: RPL17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human