To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Nitric Oxide Synthase 1 (Neuronal)
- Antigen Symbol:nNOS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Guinea pig,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:4842
- Antigen Synonyms:N-NOS|Peptidyl-cysteine S-nitrosylase NOS1|Constitutive NOS|bNOS|EC 1.14.13.39|nNOSNOS type I|brain|NOS|IHPS1|nitric oxide synthase 1 (neuronal)|Neuronal NOS
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Synthetic peptide with sequence ESKKDTDEVFSS, representing the C-terminus of the human protein (residues 1423-1434) according to NP_000611.
- Tested Applications:, Peptide Whole-Mount
- Purification:Immunogen affinity purified
- Cat. No.:102118-702
- Supplier no.:NB100-858
Specifications
About this item
The nNOS Antibody from Novus Biologicals is a goat polyclonal antibody to nNOS. This antibody reacts with human, mouse, rat, guinea pig, monkey. The nNOS Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA, Immunohistochemistry Whole-Mount.
Type: Primary
Antigen: nNOS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Guinea Pig, Monkey