To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:GABA-A R beta 3
- Antigen Symbol:GABRB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Epitope:internal region
- Gene ID:2562
- Antigen Synonyms:ECA5|MGC9051|beta 3|GABA-alpha receptor beta-2 subunit|gamma-aminobutyric acid (GABA) A receptor|GABA(A) receptor subunit beta-3|GABA(A) receptor beta-3 subunit|gamma-aminobutyric acid receptor subunit beta-3|GABAA receptor beta-3 subunit
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-EKTAKAKNDRSK corresponding to internal region according to NP_068712.1.
- Purification:Immunogen affinity purified
- Cat. No.:102157-560
- Supplier no.:NBP1-06988
Specifications
About this item
The GABA-A R beta 3 Antibody from Novus Biologicals is a goat polyclonal antibody to GABA-A R beta 3. This antibody reacts with human. The GABA-A R beta 3 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Type: Primary
Antigen: GABA-A R beta 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal region
Host: Goat
Isotype: IgG
Reactivity: Human