To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CaM Kinase II alpha
- Antigen Symbol:CAMK2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Epitope:C-PRTAQSEETRVWHR
- Gene ID:815
- Antigen Synonyms:CaM kinase II alpha subunit|CaMK-II subunit alpha|CaMK-II alpha subunit|CaM kinase II subunit alpha|calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha|CaM-kinase II alpha chain|calcium/calmodulin-dependent protein kinase II alpha-B subunit|CaMKIINalpha|EC 2.7.11|calcium/calmodulin-dependent protein kinase II alpha|CAMKAcalcium/calmodulin-dependent protein kinase type II subunit alpha|EC 2.7.11.17|KIAA0968calcium/calmodulin-dependent protein kinase type II alpha chain
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence C-PRTAQSEETRVWHR corresponding to internal region according to NP_741960.1.
- Purification:Immunogen affinity purified
- Cat. No.:102182-386
- Supplier no.:NBP1-51945
Specifications
About this item
The CaM Kinase II alpha Antibody from Novus Biologicals is a goat polyclonal antibody to CaM Kinase II alpha. This antibody reacts with human, mouse, rat. The CaM Kinase II alpha Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Type: Primary
Antigen: CaM Kinase II alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-PRTAQSEETRVWHR
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat