To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Myelin PLP
- Antigen Symbol:Myelin PLP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:0.25 ml
- Environmentally Preferable:
- Gene ID:5354
- Antigen Synonyms:HLD1|proteolipid protein 1|major myelin proteolipid protein|myelin proteolipid protein|SPG2|MMPL|lipophilin|uncomplicated|spastic paraplegia 2|PMD|PLP|PLP/DM20
- Storage Buffer:10mM Sodium PBS (0.9% isotonic, w/v, pH 7.2)
- Storage Temperature:Store at 4C in the dark.
- Concentration:0.1 mg/ml
- Immunogen:Chickens were immunized with a peptide/ keyhole limpet hemocyanin (KLH) conjugate. The synthetic peptide corresponded to a region shared between the mouse (P60202, NCBI) and human gene products [PLP1 isoform 1 (NP_000524, NCBI), PLP1 isoform 2 (NP_955772, NCBI)]. The sequence can be found between amino acid 26-75. After repeated injections into the hens, immune eggs were collected, and the IgY fractions were purified from the yolks. These IgY fractions were then affinity-purified using a peptide column, the concentration of the eluate adjusted to 100 ug/ml, and the preparation was filter-sterilized.
- Purification:Immunogen affinity purified
- Cat. No.:102105-638
- Supplier no.:NB100-1608
Chicken Polyclonal Myelin PLP Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Mouse.
Type: Primary
Antigen: Myelin PLP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity: Mouse