To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 μg
- Environmentally Preferable:
- Storage Conditions:Store at -20 to -80C. Avoid freeze-thaw cycles.
- Endotoxin Content:<0.1 ng/ug
- Gene ID:348
- Protein Synonyms:apolipoprotein E3|Apo-E|apolipoprotein E|MGC1571|late onset)
- Protein/Peptide Name:Apolipoprotein E/ApoE
- Purity:>90%, by SDS-PAGE
- Cat. No.:102224-628
- Supplier no.:NBP1-99158
Specifications
About this item
The Recombinant Human Apolipoprotein E/ApoE Protein from Novus Biologicals is derived from E. coli. The Recombinant Human Apolipoprotein E/ApoE Protein has been validated for the following applications: Western Blot, SDS-Page.