To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:TOPO2 Beta
- Antigen Symbol:TOP2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:7155
- Antigen Synonyms:EC 5.99.1.3|topoisomerase (DNA) II beta (180kD)|U937 associated antigen|topo II beta|DNA topoisomerase 2-beta|topoisomerase (DNA) II beta 180kDa|antigen MLAA-44|DNA topoisomerase II beta|topoisomerase IIb|topoisomerase II beta|DNA topoisomerase II|beta isozyme|top2beta|180 kD|TOPIIB
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 1570 and the C-terminus (residue 1621) of human DNA Topoisomerase II Beta using the numbering given in entry NP_001059.2 (GeneID 7155).
- Purification:Immunogen affinity purified
- Cat. No.:102108-936
- Supplier no.:NB100-40842
The TOP2B Antibody from Novus Biologicals is a rabbit polyclonal antibody to TOP2B. This antibody reacts with human, mouse. The TOP2B Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TOP2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse