To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:SRA Stem-Loop Interacting RNA Binding Protein
- Antigen Symbol:SLIRP
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Primates,Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:81892
- Antigen Synonyms:SLIRP|SRA stem-loop-interacting RNA-binding protein|mitochondrial|DC50|chromosome 14 open reading frame 156|PD04872
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:A synthetic peptide made to a C-terminal region of human SLIRP protein. [Entrez-Prot# AAX58600]
- Purification:Immunogen affinity purified
- Cat. No.:102121-762
- Supplier no.:NB110-37258G
Specifications
About this item
The SLIRP Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to SLIRP. This antibody reacts with human, mouse, primate. The SLIRP Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: SLIRP
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Primate