About this item
The Perilipin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Perilipin. This antibody reacts with human, mouse, rat, porcine. The Perilipin Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Perilipin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102122-040
- 102122-054
- Antigen Symbol:
- PLIN1
- PLIN1
- Antigen Name:
- Perilipin
- Perilipin
- Antigen Synonyms:
- perilipin 1|perilipin|perilipin-1|PLINLipid droplet-associated protein|PERI
- perilipin 1|perilipin|perilipin-1|PLINLipid droplet-associated protein|PERI
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Pig, Human, Rat, Mouse
- Pig, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 5346
- 5346
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- A synthetic peptide made to a region between residues 450-522 (C-terminus) of the human Perilipin protein. [Swiss-Prot# O60240]
- A synthetic peptide made to a region between residues 450-522 (C-terminus) of the human Perilipin protein. [Swiss-Prot# O60240]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1.1 mg/ml
- 1.1 mg/ml
- Storage Buffer:
- Tris-Glycine and 0.15M NaCl
- Tris-Glycine and 0.15M NaCl
- Storage Temperature:
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.