To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:COX-2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.5 ml
- Environmentally Preferable:
- Gene ID:5743
- Antigen Synonyms:hCox-2|EC 1.14.99.1|prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase andcyclooxygenase)|PGH synthase 2|COX-2|PHS II|PHS-2|Prostaglandin-endoperoxide synthase 2|EC 1.14.99|prostaglandin G/H synthase and cyclooxygenase|cyclooxygenase-2|GRIPGHS|Prostaglandin H2 synthase 2|COX2cyclooxygenase 2b|PGG/HS|prostaglandin G/H synthase 2|PGHS-2
- Storage Buffer:PBS with 2% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Immunogen:Recombinant protein corresponding to the C-terminus of rat COX2 (the exact amino acid sequence is considered to be proprietary information).
- Purification:Immunogen affinity purified
- Cat. No.:102115-634
- Supplier no.:NB100-689
Specifications
About this item
The COX-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to COX-2. This antibody reacts with human, mouse, rat. The COX-2 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:100
Type: Primary
Antigen: COX-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat