To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Troponin I Type 3 (cardiac)
- Antigen Symbol:TNNI3
- Clonality:Monoclonal
- Clone:19C7
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Fish,Pig,Rabbit,Goat,Human,Bovine,Dog,Rat,Mouse,Cat
- Western Blot:Yes
- Size:0.2 mg
- Environmentally Preferable:
- Epitope:41-49 aa
- Gene ID:7137
- Antigen Synonyms:cardiac muscle|CMD2A|RCM1|TNNC1troponin I|Cardiac troponin I|CMD1FF|cTnI|cardiac|troponin I type 3 (cardiac)|familial hypertrophic cardiomyopathy 7|troponin I|CMH7MGC116817
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C. Do not freeze.
- Immunogen:Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with free human cardiac troponin I.
- Tested Applications:Immunoassay,
- Purification:Protein A purified
- Cat. No.:102121-654
- Supplier no.:NB110-2546
Specifications
About this item
The Troponin I Type 3 (cardiac) Antibody (19C7) from Novus Biologicals is a mouse monoclonal antibody to Troponin I Type 3 (cardiac). This antibody reacts with human, mouse, rat, porcine, bovine, canine, feline, fish, goat, rabbit. The Troponin I Type 3 (cardiac) Antibody (19C7) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoassay, Sandwich ELISA.
Type: Primary
Antigen: Troponin I Type 3 (cardiac)
Clonality: Monoclonal
Clone: 19C7
Conjugation: Unconjugated
Epitope: 41-49 aa
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Canine, Feline, Fish, Goat, Rabbit