About this item
The STEP Antibody (23E5) from Novus Biologicals is a mouse monoclonal antibody to STEP. This antibody reacts with human, mouse, rat. The STEP Antibody (23E5) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
Type: Primary
Antigen: STEP
Clonality: Monoclonal
Clone: 23E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102130-198
- 102130-200
- Antigen Symbol:
- STEP
- STEP
- Antigen Name:
- Striatum-enriched protein-tyrosine phosphatase
- Striatum-enriched protein-tyrosine phosphatase
- Antigen Synonyms:
- Striatum-enriched protein-tyrosine phosphatase|FLJ14427|STEPPTPSTEP|Neural-specific protein-tyrosine phosphatase|tyrosine-protein phosphatase non-receptor type 5|protein tyrosine phosphatase|protein-tyrosine phosphatase striatum-enriched|non-receptor type 5 (striatum-enriched)|EC 3.1.3.48
- Striatum-enriched protein-tyrosine phosphatase|FLJ14427|STEPPTPSTEP|Neural-specific protein-tyrosine phosphatase|tyrosine-protein phosphatase non-receptor type 5|protein tyrosine phosphatase|protein-tyrosine phosphatase striatum-enriched|non-receptor type 5 (striatum-enriched)|EC 3.1.3.48
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 23E5
- 23E5
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 84867
- 84867
- Isotype:
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- A synthetic peptide of rat STEP46 (amino acids 57-74: MCTPGCNEEGFGYLVSPR)
- A synthetic peptide of rat STEP46 (amino acids 57-74: MCTPGCNEEGFGYLVSPR)
- Purification:
- Unpurified
- Unpurified
- Storage Buffer:
- Ascites
- Ascites
- Storage Temperature:
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.