Order Entry
United States
ContactUsLinkComponent
Thallium(I) fluoride ≥99%
Thallium(I) fluoride ≥99%
Catalog # 101776-390
Supplier:  Matrix Scientific
CAS Number:  
Thallium(I) fluoride ≥99%
Catalog # 101776-390
Supplier:  Matrix Scientific
Supplier Number:  002838-25G
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    25 g
  • Environmentally Preferable:
  • Cat. No.:
    101776-390
  • Supplier no.:
    002838-25G

Specifications

Specification Test Results

  • Boiling point:
    665°

Specification Test Results

About this item

 :
 :223.38
UN:1707
CAS Number:7789-27-7
Formula:FTl
MDL Number:MFCD00049607
Boiling Pt:645
Melting Pt:315
Density:8.36
ADR:6.1 II
Merck Index:12,09397