To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- CAS No.:7298-84-2
- Source:
- Species:
- Size:100 mg
- Environmentally Preferable:
- Protein/Peptide Name:L-Leucyl-L-Alanine
- Purity:>98.0% (by Titrimetry)
- Molecular Weight:202.25
- Color:White
- Formulation:Crystals
- Molecular Formula:C₉H₁₈N₂O₃
- Cat. No.:TCL0126-100MG
- Supplier no.:L0126-100MG
Specifications
About this item
CAS Number: 7298-84-2
MDL Number: MFCD00002641
Molecular Formula: C9H18N2O3
Molecular Weight: 202.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 19 deg (C=5, MeOH)
Storage Temperature: 0-10°C
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Leu-Ala-OH