Sodium sulfate, anhydrous, crystals USP, Ph. Eur., BP, FCC suitable for use as excipient
Catalog # 89234-940
Supplier: Avantor
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Poly drum
- Size:12 kg
- Environmentally Preferable:
- Cat. No.:89234-940
- Supplier no.:51104000120
Specifications
Specification Test Results
- Identity A) Sulfate (PhEur/BP/USP/FCC):Pass test
- Identity B) Sodium (PhEur/BP/USP/FCC):Pass test
- Identity C) Loss on drying (PhEur/BP):Pass test
- Appearance (PhEur/BP):White or almost white powder, hydroscopic.
- Solubility (PhEur/BP):Freely soluble in water.
- Assay (PhEur/BP):98.5-101.0%
- Appearance of solution (PhEur/BP):Clear and colorless.
- Acidity or alkalinity (PhEur/BP/USP):Corresponds to test
- Chlorides (PhEur/BP):450 ppm Max.
- Calcium (PhEur/BP):450 ppm Max.
- Iron (PhEur/BP):90 ppm Max.
- Loss on drying (PhEur/BP/USP):0.5% Max.
- Assay (USP):99% Min.
- Chlorides (USP):0.02% Min.
- Heavy metals (USP):0.001% Min.
- Appearance (FCC):Colorless crystals or as a fine, white, crystalline powder.
- Solubility (FCC):Freely soluble in water and practically insoluble in alcohol.
- Assay (FCC):99.0 – 100.5 %
- Loss on drying (FCC):1% Max.
- Lead (FCC):2 mg/kg Max.
- Selenium (FCC):0.003% Max.
Specification Test Results
About this item
Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt
:
:142.04 g/mol
Boiling Pt:1429
Melting Pt:888
Density:2.7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-820-9
CAS Number:7757-82-6
Formula:Na₂SO₄
MDL Number:MFCD00003504
Merck Index:12,08829