Specifications
- Description:Protein Precipitation Kit
- Size:Kit
- Environmentally Preferable:
- Cat. No.:71005-978
- Supplier no.:1B1301-KIT
Specifications
About this item
VWR® Protein Precipitation Kit utilizes a simple trichloroacetic acid (TCA) based method to precipitate proteins in complex mixtures
- Removal of interfering salts, detergents, and buffers
- Reproducible and Consistent Results
Only protein in the sample is pelleted, while interfering salts and surfactants remain in the supernatant. A gentle acetone wash removes TCA and co-precipitants allowing the end-user to recover the protein in a small volume of buffer. Kit contains: Protein Precipitation Kit, Solution A, 25 mL, Protein Precipitation Kit, Solution B, 250 mL, and Wash Buffer, 125 mL.