Zinc sulfate heptahydrate ≥99%, crystals, BAKER ANALYZED® ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
CAS Number:7446-20-0
:
Formula:ZnSO₄·7H₂O
MDL Number:MFCD00149894
Density:1.97 g/cm³ (20 °C)
Molecular Weight:287.54 g/mol
Melting Pt:100
ADR:9 III
Merck Index:12,10293
:Management of Change (MOC) category = NI
EINECS:231-793-3
UN:3077
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- ACS - Assay (ZnSO₄·7H₂O):99.0 - 103.0 %
- ACS - Insoluble Matter:≤0.005 %
- ACS - pH of 5% Solution at 25°C:4.4 - 6.0
- ACS - Nitrate (NO₃):≤0.002 %
- ACS - Ammonium (NH₄):≤0.001 %
- ACS - Calcium (Ca):≤0.005 %
- ACS - Lead (Pb):≤0.003 %
- ACS - Magnesium (Mg):≤0.005 %
- ACS - Potassium (K):≤0.01 %
- ACS - Sodium (Na):≤0.05 %
- Chloride (Cl):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- Trace Impurities - Manganese (Mn):≤2.000 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number4382 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT4382-5
Product specification
Frequently Bought Together