About this item
These kits are an excellent choice for sample collection on large surfaces or equipment parts
The benefits of foam-tipped swabs are the large tip, which increases sample and elution counts, and the rigid paddle-like design which allows for the firm and even application of pressure necessary for adequate sample collection. To use, simply remove Puritan's EnviroMax® swab from the vial, collect the sample, and return the swab to the vial.
Shelf life is 12 months. Made in USA.
The oversized swab consists of a rigid paddle inside a foam tip. The swab tip is made out of a medical-grade polyurethane foam and is securely attached to a polypropylene handle. Tips are pointed for use in hard-to-reach areas such as drains.