ContactUsLinkComponent
Nitrate and Nitrite Test Strips, Hach
Nitrate and Nitrite Test Strips, Hach
Catalog # 97026-442
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
undefined
Nitrate and Nitrite Test Strips, Hach
Catalog # 97026-442
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Environmentally Preferable:
  • Cat. No.:
    97026-442
  • Range:
    Nitrate 0 - 50 mg/L, Nitrite 0 - 3.0 mg/L
  • Supplier no.:
    2745425
  • No. of tests:
    25
Specifications
About this item
Excessive amounts of nitrate or nitrite in water can cause infant death, adult illness, and can produce spontaneous abortion in cows. Some wells contain high levels of nitrate. When water contains high nitrate levels, nitrites are often present in low concentration.
With Hach Water Quality Test Strips, technicians in the field can test many samples in only a few minutes and make immediate evaluations on-site
Easy to use, disposable, and inexpensive. A great way to obtain quick, quantitative answers in the field or in the lab.
No measuring, set-up, clean-up, or chemical handling are necessary. Hach test strips are also used in laboratories all over the world for pre-test screening-to detect the presence of materials that might interfere with lab testing. Contains 25 tests.