SP Bel-Art Enviro-Safe® General Purpose Liquid In Glass Thermometer, Bel-Art Products, a part of SP
Supplier: Avantor
Certificates
About this item
Earth friendly, EnviroKleen™ certified thermometers feature green, non-toxic, biodegradable, Enviro-Safe® liquid against lead-free, white back glass
- Meet government, education, and industry mandates to purchase “Green” products
- Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries
- Manufactured and tested by our USA quality team
- Individually serialized
Include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Certifications: Traceable to NIST.
Related Information
AT-VWR Thermometer FAQs
Datasheet
ED EnviroKleen Certification
Additional Product Details
Frequently Bought Together