Order Entry
United States
ContactUsLinkComponent
Distilled Water
Distilled Water
  470200-788
 :  Avantor
 :  
Distilled Water
  470200-788
 :  Avantor
 :  

 

  • Description:
    Distilled Water
  • Size:
    1 Gal
  • Environmentally Preferable:
  • Cat. No.:
    470200-788
  • Grade:
    Laboratory
  • Supplier no.:
    887005

 

 

CAS Number: 7732-18-5
Formula: H2O
Density: 1 g/mL
Boiling and Freezing Point: 100°C and 0°C

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.