Variable Inductor
470128-394
: Wards
- Description:Variable Inductor
- Environmentally Preferable:
- Cat. No.:470128-394
- Supplier no.:PH0845VIC
Adjustable for Precision
- Compatible with 4 mm safety sockets.
- Includes a carrying handle for easy portability.
The screw adjustment enameled wire winding built into this unit allows precise measurements up to 1.3 H. Soft iron core displacement includes a scale mark and quick advancement worm.