Ward's® Live Termite (Zootermopsis sp.)
470180-354
- Description:Termites
- Environmentally Preferable:
- Cat. No.:470180-354
- Supplier no.:WARD470180-354
This species of termite is very large, and you can easily observe the distinct differences of the several roles that exist in their eusocial colonies.
- Exhibit "Caste" system
- Excellent model for symbiosis
- Extremely easy to maintain in the laboratory or classroom
- No reproductive termites are sent (we do not send queens)
One of the two different genera of termites available at Ward's, Zootermopsis are known as dampwood termites. They are commonly located in the Pacific northwest and southern British Columbia where they make their homes in rotting logs and other dark, moist wooden areas. They have a symbiotic relationship with a flagellate protozoa in their gut that helps them digest the cellulose in the wood.
: These products contain living or perishable materials. No reproductive termites are sent with the culture. We do not send queens.
: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Ward's holds valid USDA permits allowing us to ship to most continental US states. Please call Customer Service for permit details.
Ward's holds valid USDA permits allowing us to ship to most continental US states. Please call Customer Service for permit details.