Taenia pisiformis, Gravid Proglottid Slide
Catalog # 470182-110
Supplier: Avantor
Specifications
- Description:Taenia pisiformis, Gravid Proglottid
- Environmentally Preferable:
- Cat. No.:470182-110
- Stain:Schneider's and Fast Green
- Supplier no.:WARD470182-110
- Orientation:Whole mount
Specifications
About this item
Endoparasitic Tapeworm
- Identifying features clearly distinguishable
- Rigorous quality control standards
Gravid proglottid of Taenia pisiformis, showing eggs.