ContactUsLinkComponent
Ward's® Simple Stains for Bacteria Lab Activity
Ward's® Simple Stains for Bacteria Lab Activity
Catalog # 470007-174
Supplier:  Avantor
undefined
Ward's® Simple Stains for Bacteria Lab Activity
Catalog # 470007-174
Supplier:  Avantor
Specifications
  • Description:
    Simple Stains for Bacteria Kit
  • Environmentally Preferable:
  • No. of lab groups:
    30
  • Cat. No.:
    470007-174
Specifications
About this item
Practice Staining Techniques to Identify Bacterial Shapes and Spores
  • Employ multiple staining techniques
  • Materials for 30 lab groups
  • Time Required: ~75 minutes
Cocci, bacilli, and spirilliform types of bacteria are easy to see with the simple staining techniques employed in this investigation. Crystal violet and methylene blue stains reveal cellular morphology; in addition, carbol fuchsin stains spores, helping to illustrate how their formation protects bacteria when the organism is under stress. Tube cultures: Aquaspirillum serpens (spirillum), Bacillus cereus (bacillus), and Staphylococcus epidermidis (cocci). The set includes enough materials for 30 students, teacher’s guide, and instructions.
Canada: Containment Level 2 – PHAC License required.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.