ContactUsLinkComponent
Sickle Cell Anemia
Sickle Cell Anemia
Catalog # 470177-656
Supplier:  Avantor
undefined
Sickle Cell Anemia
Catalog # 470177-656
Supplier:  Avantor
Specifications
  • Description:
    Sickle Cell Anemia, smear
  • Environmentally Preferable:
  • Cat. No.:
    470177-656
  • Stain:
    Giemsa
  • Supplier no.:
    WARD470177-656
  • Orientation:
    Smear
Specifications
About this item
Smear, giemsa stain
  • All identifying features are clearly distinguishable
  • Rigorous quality control standards
Smears of human peripheral blood collected from a patient with Sickle Cell Anemia. Presence of crescent-shaped red blood cells will vary from patient to patient, thus from slide-to-slide.