Specifications
- Description:Ward's® Chemistry Beautiful Balancing Buffers Demonstration
- Environmentally Preferable:
- No. of lab groups:5
- Cat. No.:470016-840
- Includes:Instructions
- Supplier no.:WARD470016-840
Specifications
About this item
How do buffers really work?
- Materials are provided to repeat the demonstration five times
Add a strong acid or a strong base to a non-buffered solution and demonstrate, using a colorful pH indicator, that the pH quickly changes. Now add the same acid and base to a buffer solution and watch how the pH stays beautifully balanced. This demonstration allows you to teach what components are required for every buffer and then apply this to a common household buffer, Alka-Seltzer®. Summarize the lesson by testing the buffering ability of an Alka-Seltzer® solution and in another beautiful balancing act.
Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.