Order Entry
United States
ContactUsLinkComponent
Rat Skull
Rat Skull
Catalog # 470027-960
Supplier:  Avantor
undefined
Rat Skull
Catalog # 470027-960
Supplier:  Avantor

Specifications

  • Description:
    Rat Skull
  • Height:
    1"
  • Length:
    2"
  • Environmentally Preferable:
  • Cat. No.:
    470027-960
  • Width:
    1"
  • Supplier no.:
    WARD470027-960
  • No. of parts:
    1

Specifications

About this item