ContactUsLinkComponent
Fundamentals of Physics Pendulum Kit
Fundamentals of Physics Pendulum Kit
Catalog # 470016-330
Supplier:  Wards
undefined
Fundamentals of Physics Pendulum Kit
Catalog # 470016-330
Supplier:  Wards
Supplier Number:  PND08
Specifications
  • Description:
    Fundamentals of physics pendulum kit
  • Dimensions:
    9L x 7 3/4W x 2 3/4"H
  • Environmentally Preferable:
  • Cat. No.:
    470016-330
  • Supplier no.:
    PND08
Specifications
About this item
This Pendulum kit is an excellent model for investigating energy.
  • Large-Scale Protractor
  • Easy-To-Change Masses Included
  • Teacher's Manual Included
  • Adjustable String Length
  • Students Learn How Different Factors Affect a Pendulum's Period
Users can adjust string length and the mass of the pendulum. A large protractor makes angle measurements easy to read, and the photogate system can be attached at the bottom of the swing arc to record the period.
Students can investigate how the angle of release, mass, and string length affect the period of a pendulum.
Accessories information:
The FUN stand and fundamentals of physics photogate system are required, but available separately.
Ordering information:
This kit includes a protractor backboard, string, pendulum bob and weights, mounting screw, and activity guide.
Caution:
Small parts can cause a choking hazard. This product is not for children under three years.