ContactUsLinkComponent
Patella, Trochophore Larva Slide
Patella, Trochophore Larva Slide
Catalog # 470182-332
Supplier:  Avantor
undefined
Patella, Trochophore Larva Slide
Catalog # 470182-332
Supplier:  Avantor
Specifications
  • Description:
    Patella, Trochophore Larva
  • Environmentally Preferable:
  • Cat. No.:
    470182-332
  • Stain:
    Schneider's and Fast Green
  • Supplier no.:
    WARD470182-332
  • Orientation:
    Whole mount
Specifications
About this item
Genus of Sea Snail
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Marine limpet. Pear-shaped free-swimming ciliated larva with a complete digestive tract.