To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:Olive Oil
- Size:500 mL
- Environmentally Preferable:
- Chemical/physical specifications:Solution
- Cat. No.:470301-908
- Grade:Laboratory
- Supplier no.:OX0020-500ML
CAS Number: 8001-25-0
Density (g/mL): 0.910-0.915
Boiling Point (°C): will not boil @ 760 mm
Freezing Point (°C): -6
Solubility: Carbon Disulfide, Chloroform, Ether
Synonyms: Sweet Oil
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.