Moth and Butterfly Antennae Slide
Catalog # 470176-942
Supplier: Avantor
Specifications
- Description:Moth & Butterfly Antennae
- Environmentally Preferable:
- Cat. No.:470176-942
- Stain:Unstained
- Supplier no.:WARD470176-942
- Orientation:Whole mount
Specifications
About this item
Illustrates Differences Between the Two Structures
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows comparative differences between moth and butterfly antennae. Unstained.