Ward's® Live Mixed Green Algae
Supplier: Avantor
Certificates
About this item
One jar containing a mixture of 6 different genera of algae. Each algae is cultured individually in Ward's laboratories, and mixed together immediately prior to shipment to ensure an adequate supply of each organism.
- Teach your students about algal diversity using our specially prepared, "real-life" combinations
- Includes dichotomous key to help your students identify each organism by key structures
The world is teeming with a wide variety of micro life, making studying samples of pond water complicated especially for the beginning student. To lessen the confusion, WARD’S has prepared different culture mixes that reflect protist combinations that occur in the real world, yet eliminates the hundreds of other life-forms found in a collected sample.
Ordering information: These products contain living or perishable materials, please specify a delivery date during checkout.
Delivery: Living specimens will ship via overnight or 2nd day delivery, with delivery available Tuesday through Friday
Packaging: Cultures include organisms from the following genera (species are not specified for these organisms): Chlamydomonas, Oscillatoria, Pandorina, Scenedesmus, Spirogyra and Ulothrix.
Frequently Bought Together