Mini ProBath Waterbath
Catalog # 470013-450
Supplier: Revolutionary Science
Specifications
- Capacity:0,5 L
- Temperature Range:Ambient...100 °C
- Description:Mini Pro Small Waterbath
- Environmentally Preferable:
- Cat. No.:470013-450
- Supplier no.:RS-PB-50
- Ø×H:8×4½"
Specifications
About this item
A fraction of the cost of other water baths, the Mini ProBath Water bath is as versatile as larger units.
- Compact Model, 0.5 L
- Diameter: 8"
- Height: 4½"
- Ideal for Warming Agar
Ideal for warming agar, the water bath can also be used to create a steam bath or keep media warm.
Its temperature range is ambient–100°C.
Ordering information: This water bath includes a heating-indicator light, temperature regulator control knob, and transparent polymer cover.