Milk of Magnesia
Catalog # 470150-466
Supplier: Avantor
CAS Number:
Specifications
- Description:Milk of Magnesia
- Size:12 oz
- Environmentally Preferable:
- Chemical/physical specifications:Solution
- Cat. No.:470150-466
- Grade:Laboratory
- Supplier no.:809327
Specifications
About this item
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.