Specifications
- Description:Pre-cleaned slides
- L×W:3x1"
- Environmentally Preferable:
- Cat. No.:470151-068
- Supplier no.:630-004T
Specifications
About this item
Quality microscope slides to make your own slide preparations.
- High quality slide making materials
- Dust free and ready to use
Pre-cleaned, premium grade microscope slides. Carefully selected to be non-corrosive and free of flaws, chemicals, and dust. Optically flat with ground edges and annealed to minimize chipping.