ContactUsLinkComponent
Low Temperature Thermometers in °F/°C
Low Temperature Thermometers in °F/°C
  470111-942
 :  Echo
undefined
Low Temperature Thermometers in °F/°C
  470111-942
 :  Echo
 :  451
  • Description:
    Low Temperature Thermometers in °C/°F or °C, Scale: Dual, Range: -30 to 50 °C, -20 to 120 °F
  • Environmentally Preferable:
  • Cat. No.:
    470111-942
  • Supplier no.:
    451
Low cost thermometers for beginning students.
Glass tubes with red alcohol mounted on 1-1/2" x 6" plastic backs with indelible scales. Graduated: 2°. Package of 15.