ContactUsLinkComponent
Fundamentals of Physics Loop Track
Catalog # 470014-340
Supplier:  Wards
undefined
Fundamentals of Physics Loop Track
Catalog # 470014-340
Supplier:  Wards
Supplier Number:  LPL008
Specifications
  • Description:
    FUNdamentals of Physics Loop Track
  • Environmentally Preferable:
  • Cat. No.:
    470014-340
  • Supplier no.:
    LPL008
Specifications
About this item
The versatile Loop Track helps students learn about acceleration and conservation of energy.
  • Activity Manual Included
  • Students Investigate Acceleration
  • Activity: Observing a Marble's Minimum Velocity
  • Topics Include Acceleration and Conservation of Energy
With this comparable experiment, students investigate how a roller coaster car can stay on the track when it travels in a large, circular loop.
Use a straight, 80 cm long portion of the track with a marble to investigate acceleration. The circular portion lets students experimentally determine the minimum velocity the marble needs to make one full trip around the loop.
Accessories information: The Photogate System, available separately, allows for simple calculations when used with the Loop Track.
Ordering information: This kit includes an activity manual. The Fun Stand is required but available separately.