ContactUsLinkComponent
Vernier® Logger Pro® Software
Vernier® Logger Pro® Software
Catalog # 470120-710
Supplier:  Avantor
undefined
Vernier® Logger Pro® Software
Catalog # 470120-710
Supplier:  Avantor
Specifications
  • Description:
    Logger Pro CD-ROM, Site License
  • Environmentally Preferable:
  • Cat. No.:
    470120-710
  • Supplier no.:
    LP-C
Specifications
About this item
You can collect, graph, and analyze data in real-time with a LabPro.
  • Site license includes home computers of students and faculty
  • No need to count computers to satisfy licensing
  • Software can be downloaded anytime
  • Students can receive software from online accounts
  • Free logger Pro 3 updates
  • Dimensions: 121×53×39 mm
In addition to collecting and analyzing sensor data, the Vernier Logger Pro software analyzes videos captured with a DV camcorder or imported from a digital camera. With Logger Pro, you can capture and analyze video with and without sensors.
A single copy has a site license for every computer in your school or college department and teacher's and student's home computers. It is for educational purposes only.
Ordering information: Software requires Windows XP, Vista (32 or 64-bit), Windows 7 (32 or 64-bit), or Mac OS X (10.4, 10.5, 10.6).