ContactUsLinkComponent
Labnet Vortex Mixer
Labnet Vortex Mixer
Catalog # 470127-070
undefined
Labnet Vortex Mixer
Catalog # 470127-070
Supplier Number:  S0200
Specifications
  • Description:
    Labnet Vortex Mixer
  • Environmentally Preferable:
  • Cat. No.:
    470127-070
  • Supplier no.:
    S0200
Specifications
About this item
Powerful and Reliable
Quickly mix and re-suspend your samples with this powerful yet compact laboratory mixer. The standard combi-cup accepts a variety of tube sizes up to 50 ml. and includes multiple accessory heads to expand its capability for mixing several tubes at once. It's powerful circular orbit provides effective vortexing at any speed, and a balanced drive system prevents the mixer from "walking" across your countertop.