Jejunum, Human Slide
Catalog # 470183-054
Supplier: Avantor
Specifications
- Description:Jejunum, Human Slide
- Environmentally Preferable:
- Cat. No.:470183-054
- Stain:Hematoxylin and Eosin
- Supplier no.:WARD470183-054
- Orientation:Section
Specifications
About this item
Midsection of intestine
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Absorbs nutrients. Numerous circularly arranged folds of the mucosa and submucosa, known as plicae circulares.