Ward's® Live Sarcophaga Bullata Fly Larvae and Pupae
: Avantor
Sarcophaga bullata are gray flesh flies. These flies are reared on decaying flesh and pupate quickly, within 5 to 7 days from birth, to illustrate complete metamorphosis in Diptera.
The (uninfected) pupae can serve as the host organism for emerging Nasonia larvae. The pupae can be stored under refrigeration for several weeks, effectively halting their life cycle and assuring a fresh supply of food for the Nasonia larvae.
: Please specify a delivery date during checkout.
To request a specific instar, please contact us ten days in advance.
To request a specific instar, please contact us ten days in advance.
: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
: A large, vigorous culture is shipped in a shatterproof plastic vial.
Frequently Bought Together