Echinococcus sp., Hydatid Cyst Slide
Catalog # 470182-090
Supplier: Avantor
Specifications
- Description:Echinococcus sp., Hydatid Cyst
- Environmentally Preferable:
- Cat. No.:470182-090
- Stain:Hematoxylin and triosin
- Supplier no.:WARD470182-090
- Orientation:Section
Specifications
About this item
Intestinal Parasite
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows the relationship of the brood capsule, scolices, and cyst wall.