Order Entry
United States
ContactUsLinkComponent
DNA-RNA-Protein Synthesis Model
DNA-RNA-Protein Synthesis Model
Catalog # 470007-366
Supplier:  The Science Source
undefined
DNA-RNA-Protein Synthesis Model
Catalog # 470007-366
Supplier:  The Science Source
Supplier Number:  B118

Specifications

  • Description:
    DNA-RNA-Protein Synthesis Kit
  • Environmentally Preferable:
  • Cat. No.:
    470007-366
  • Supplier no.:
    B118

Specifications

About this item

Designed To Show Protein Synthesis In A Single Model

  • Demonstrate Protein Synthesis In One Model
  • Easily Assembled
  • Stores Easily
  • H x L x W: 18 cm x 61 cm x 51 cm

Demonstrate how all the nucleic acid elements relate to each other in the production of amino acids using this comprehensive molecular kit. The easily assembled model consists of 12 DNA bases, 12 mRNA, 12 tRNA bases, four amino acids, and six molded rubber strands representing phosphate, which all mount on a wooden frame. Includes a leatherette carrying/storage case and an illustrated instruction manual.