Cycloid, Scale Slide
Catalog # 470181-718
Supplier: Avantor
Specifications
- Description:Cycloid, Scale
- Environmentally Preferable:
- Cat. No.:470181-718
- Supplier no.:WARD470181-718
- Orientation:Whole mount
Specifications
About this item
Scale Type of Fish
- Identifying features clearly distinguishable
- Rigorous quality control standards
Stained. Isolated scale showing characteristic smooth curve of distal border.