Crepidula, Veliger Larva Slide
Catalog # 470182-334
Supplier: Avantor
Specifications
- Description:Crepidula, Veliger Larva
- Environmentally Preferable:
- Cat. No.:470182-334
- Stain:Schneider's and Fast Green
- Supplier no.:WARD470182-334
- Orientation:Whole mount
Specifications
About this item
Free-swimming Larva
- Identifying features clearly distinguishable
- Rigorous quality control standards
Larva develops from a trochophore and has beginnings of foot, mantle, shell.