Clostridium tetani Slide
Catalog # 470180-896
Supplier: Avantor
Specifications
- Description:Clostridium tetani,gram (+)
- Environmentally Preferable:
- Cat. No.:470180-896
- Stain:Gram (+)
- Supplier no.:WARD470180-896
- Orientation:Smear
Specifications
About this item
Pathogenic bacteria
- Identifying features clearly distinguishable
- Rigorous quality control standards
Rods. Spores round and terminal. Agent of tetanus in humans.