ContactUsLinkComponent
Chromatography Paper Strips
  470004-492
undefined
Chromatography Paper Strips
  470004-492
 :  CHROM-50-6X75
  • Description:
    Chromatography Paper Strips, Pack of 50
  • Size:
    6 x ¾"
  • Environmentally Preferable:
  • Cat. No.:
    470004-492
  • Supplier no.:
    CHROM-50-6X75
Perfect for your paper chromatography needs.
  • Package of 50
This chromatography paper is supplied in strips rather than rolls that are apt to stretch in order to assure the most even distribution of material being separated for more accurate results.