Specifications
- Description:Chemical Properties Lab Activity
- Environmentally Preferable:
- No. of lab groups:6
- Cat. No.:470002-482
- Supplier no.:13-104
Specifications
About this item
Test Chemical Properties of Seven Different Substances
- Ideal for Grades 9-12
- Chemical Properties in Microscale
Students test the chemical properties of seven different solutions in every combination and identify chemical properties by observing the reactions. Evidence of chemical reactions observed include gas production, color changes, and formation of precipitates. After becoming familiar with the chemical properties of the solutions, students use the information to identify any of four unknown solutions. The kit includes all required chemicals, reaction plates, reproducible worksheets an student procedures.