Casio FX-300MS Plus Scientific Calculator
470143-884
: Avantor
- Description:Casio calculator FX-300MS Plus
- Environmentally Preferable:
- Cat. No.:470143-884
- Supplier no.:FX-300MS
Approved for use on College Entrance Exams, including the SAT and PSAT/NMSQT.
- Approved for college entrance exams
- Features 229 functions
- two-line display
This calculator features 229 built-in functions, including fractions, statistics, standard deviation, linear regression, trigonometry, and polar-rectangular conversions. A fraction key is also included for easy entry and calculation of fractions in addition to quick conversions of fractions to decimals. Other features include a 10 digit, two-line display;random number generator; automatic power down; and protective hard case. Operates on solar and battery power.